Organic reference standard, Chloroprene solution, certified reference material, Supelco®
Catalog # 861145.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml in methanol
- Pk:1
- Standards:Certified reference material
- Description:Chloroprene solution
- Storage temperature:2...8 °C
- Description:Chloroprene solution
- Pk Info:1 ml/pack
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.