VISOCOLOR® Colorimetric Test, Hydrazine
About this item
Reagents for the determination of hydrazine in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
2 Options Available Below
Product Details & Documents
Reagents for the determination of hydrazine in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- MANA931030
- MANA931230
- Model:
- MANA931030
- MANA931230
- Platform:
- Colorimetric test kits
- Colorimetric test kits
- Storage temperature:
- 15...25 °C
- 15...25 °C
- Shelf life (months):
- 12
- 12
- Weight:
- 237 g (Including packaging)
- 174 g (Including packaging)
Specifications
Product Family Options
Test kits
Product Information
- ContentsRangeChemical methodFor unitDimensionsTests per kitPkAvailabilityPrice
- MANA931030VISOCOLOR® ECO Hydrazine, test no. 5-30VISOCOLOR® ECO Hydrazine, test no. 5-30Reagents for 130 tests, color chart and accessories in a box0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 mg/L (N₂H₄); 0,05 - 0,40 mg/L (N₂H₄)4-DimethylaminobenzaldehydePF-12Plus;Advance;Visual168×112×40 mm (Including packaging)1301Log in for availability1 * 1 UN98,50 €Specifications:
More
Specifications
Cat. No.MANA931030ModelMANA931030PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)12Weight237 g (Including packaging)
Refill packs
Product Information
- ContentsRangeChemical methodFor unitDimensionsTests per kitPkAvailabilityPrice
- MANA931230VISOCOLOR® ECO Hydrazine, refill pack, test no. 5-30VISOCOLOR® ECO Hydrazine, refill pack, test no. 5-30Reagents for 130 tests in a box0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 mg/L (N₂H₄); 0,05 - 0,40 mg/L (N₂H₄)4-DimethylaminobenzaldehydePF-12Plus;Advance;Visual170×115×70 mm (Including packaging)1301Log in for availability1 * 1 UN71,90 €Specifications:
More
Specifications
Cat. No.MANA931230ModelMANA931230PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)12Weight174 g (Including packaging)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.



