Order Entry
Spain
ContactUsLinkComponent
Photometric rapid tests, fluoride, VISOcOLOR®
Photometric rapid tests, fluoride, VISOcOLOR®
Catalog # MANA931227
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Photometric rapid tests, fluoride, VISOcOLOR®
Catalog # MANA931227
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    SPADNS
  • Pk:
    1 KIT
  • Description:
    Fluoride, VISOCOLOR® ECO
  • For unit:
    Photometer PF-12
  • Tests per kit:
    150
  • Range:
    0.1 - 2.0 mg/l

Specifications

About this item

The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.

  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges

The test is delivered without colour scale and can be evaluated only with the photometers PF-12.

Normally, the content of fluoride in surface and ground water is lower than 1 mg/L. Permanent consumption of water containing more than 2 mg/L fluoride can cause a drinking water fluorese (a stained discolouration of the tooth enamel). On the other hand a content of fluoride lower than 0,5 mg/L could lead to an increasing risk of caries. The optimal content of fluoride in drinking water is about 1 mg/L. To avoid problems with caries in some countries the content of fluoride in drinking water is artificially increased. However, WHO and EC drinking water regulations recommend a threshold value of 1,5 mg/L F⁻.

In some European countries, the threshold value for mineral water is also 1,5 mg/L F⁻. For the suitability of water for the preparation of baby food a threshold value of 0,7 mg/L F⁻ is recommended. Natural mineral waters with a fluoride content of more than 5 mg/L must be labelled with a warning.