Order Entry
Spain
ContactUsLinkComponent
Arsenic test papers, qualitative
Arsenic test papers, qualitative
Catalog # MANA90762
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Arsenic test papers, qualitative
Catalog # MANA90762
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Mercuric bromide
  • Pk:
    200
  • Description:
    Arsenic, arsine (As, AsH3)

Specifications

About this item

This test paper allows the easy detection of arsine (AsH₃) in the gas phase. Arsenic in solutions has to be converted into AsH₃ with Zn/acid and purged from the solution. The Arsine is detected directly at the boundary layer between water and air.

  • Colour change: White to brown-black

Arsenic test paper is used for the easy determination of arsenic in grape must and wine. For the determination of Arsine in compartment air the test paper is moistened with acetic anhydride.

Limit of sensitivity: 0,5 µg As

Delivery: Pack of 200 strips 20×70 mm.