Flow-through cells
About this item
Flow-through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell.
- Outside dimension: 35×12,5×12,5 mm for Quartz SUPRASIL cell with 1 and 10 mm light path
- Outside dimension: 45×12,5×52,5 mm for Quartz SUPRASIL cell with 50 mm light path
- Each pack contains one cell with two clear windows
The cells are compact with two screw connectors, M6×1 and 500 mm length FEP tubing of 1,9 mm O.Ø. and 1,1 mm I.Ø.
5 Options Available Below
Product Details & Documents
Flow-through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell.
- Outside dimension: 35×12,5×12,5 mm for Quartz SUPRASIL cell with 1 and 10 mm light path
- Outside dimension: 45×12,5×52,5 mm for Quartz SUPRASIL cell with 50 mm light path
- Each pack contains one cell with two clear windows
The cells are compact with two screw connectors, M6×1 and 500 mm length FEP tubing of 1,9 mm O.Ø. and 1,1 mm I.Ø.
Product Family Options
Product Information
- Centre heightVolumePath lengthAvailabilityPrice
- PERCB0631151Quartz SUPRASIL (FIAS) cell (Ø 1,5 mm aperture)Quartz SUPRASIL (FIAS) cell (Ø 1,5 mm aperture)15 mm18 µl10 mmLog in for availability1 * 1 UN1 170,00 €
- PERCB0631090Quartz SUPRASIL cell (Ø 2 mm aperture)
- PERCB0631087Quartz SUPRASIL cell (8×2 mm aperture)
- PERCB0631088Quartz SUPRASIL cell (Ø 3 mm aperture)
- PERCB0631084Quartz SUPRASIL cell (11×3,5 mm aperture)