

Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
:67-66-3
:
:CHCl₃
:MFCD00000826
:119.38 g/mol
:61 °C (1.013 hPa)
:–61.3 °C
:1.485 g/cm³ (20 °C)
:Ambient
:200-663-8
:6.1,III
:Stabilised with amylene
:1888
:GHS07
:GHS08
- Assay (GC):Min. 99.80 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as HCl):Max. 0.001 %
- Absorbance at 250 nm:Max. 1.00
- Absorbance at 260 nm:Max. 0.30
- Absorbance 280 nm:Max. 0.005
- IR spectrum:complying
- Vapor Density:4.1 (vs air)
- Refractive Index:n20/D 1.445 (lit.)
Frequently Bought Together