Order Entry
Spain
ContactUsLinkComponent
Perchloric acid ~60% ACS, Fluka™
Perchloric acid ~60% ACS, Fluka™
Certificates
undefined
Perchloric acid ~60% ACS, Fluka™

Specification Test Results

  • Assay:
    60.0 - 62.0 %
  • Residue on ignition:
    Max. 0.003 %
  • Iron (Fe):
    Max. 1 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Phosphate, silicate (as SiO₂):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.001 %
  • Total N:
    Max. 0.001 %
  • APHA:
    Max. 10
  • Ignition Residue:
    ≤0.003%

Specification Test Results

Related Information

Frequently Bought Together