Order Entry
Spain
ContactUsLinkComponent
Barium perchlorate 0.005 M Ba(ClO4)2 in 2-propanol/water volumetric solution, Fluka™
 
 
Barium perchlorate 0.005 M Ba(ClO4)2 in 2-propanol/water volumetric solution, Fluka™
  35053-1L
 :  
 
 
Barium perchlorate 0.005 M Ba(ClO4)2 in 2-propanol/water volumetric solution, Fluka™
  35053-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Factor:
    0.995 - 1.005

 

 

 :
 :336.23 g/mol
 :0.908 g/cm³ (20 °C)
 :21 °C
 :Ambient
 :1219
 :3,II
 :GHS07
 :GHS02
 :13465-95-7
 :Ba(ClO₄)₂
 :MFCD00003440