Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silicone elastomer
Silicone elastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Silicone elastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Silicone elastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Catalog # DOWC42417.2
Supplier:
DOW CORNING
CAS Number:
Expand image
Silicone elastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Catalog # DOWC42417.2
Supplier:
DOW CORNING
CAS Number:
Specifications
Pk:
1
Seq:
4135
Pk Info:
7,2 kg kit
Specifications
About this item
Product Details
:
Product Details