Order Entry
Spain
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator, for COD determination
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator, for COD determination
Catalog # HACH181233
Supplier:  Hach
CAS Number:  
undefined
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator, for COD determination
Catalog # HACH181233
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Dropping bottle
  • Pk:
    29 mL
  • Seq:
    2335

Specifications

About this item

 :
 :692.54 g/mol
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428