Supelco® TraceCERT® Organic Reference Standard, PAH Calibration Mix
Catalog # SUPLCRM47940
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 µg/ml each component in acetonitrile
- Pk:1 mL
- Standards:TraceCERT®, certified reference material
- Description:PAH Calibration mix
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319 - H412
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Polynuclear Aromatic Hydrocarbons Mix
- Flash point:2,0 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P273 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312