Blood sampling system, blood gas Monovette®
Supplier: SARSTEDT
Certificates
About this item
The blood gas Monovette® is available in 1 ml and 2 ml options for venous and arterial blood collection.
The Ca²⁺ balanced heparin is also suitable for determining electrolytes.
Frequently Bought Together