Order Entry
Spain
ContactUsLinkComponent
Supelco® Organic Reference Standard, DBA-d₉ Monomers Internal Standard Mix
Supelco® Organic Reference Standard, DBA-d₉ Monomers Internal Standard Mix
Catalog # CRM40570
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, DBA-d₉ Monomers Internal Standard Mix
Catalog # CRM40570
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    In acetone (varied concentration)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    DBA-d₉ Monomers internal standard mix
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H319 - H336
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –17,0 °C – Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P305 + P351 + P338