Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, sunset yellow (E110), Dr Ehrenstorfer®
Organic reference standard, sunset yellow (E110), Dr Ehrenstorfer®
  EHERC17048000
 :  
Organic reference standard, sunset yellow (E110), Dr Ehrenstorfer®
  EHERC17048000
 :  

 

  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    2783-94-0
  • Description:
    Sunset yellow
  • Formula:
    C₁₆H₁₀N₂O₇S₂·2Na
  • Description:
    Sunset yellow

 

 

 : Accredited to ISO/IEC 17025.