Order Entry
Spain
ContactUsLinkComponent
UV exposure screen for myECL™ imager, Pierce
UV exposure screen for myECL™ imager, Pierce
  PIER62244
UV exposure screen for myECL™ imager, Pierce
  PIER62244
  • Description:
    UV exposure screen for myECL™ imager, 245×195×3,2 mm
  • Pk:
    1
This clear plastic sample screen is used to cover the transilluminator on the myECL™ imager. It is a replacement for the accessory used on the transilluminator for acquiring images in the UV mode.