Order Entry
Spain
ContactUsLinkComponent
Chemiluminescence exposure screen for myECL™ imager
  PIER62242
Chemiluminescence exposure screen for myECL™ imager
  PIER62242
  • Description:
    Chemiluminescence exposure screen for myECL™ imager, 245×195×3,2 mm
  • Pk:
    1
This black plastic sample screen is used to cover the transilluminator on the myECL™ imager.
This has been used as a replacement for the accessory used on the transilluminator for acquiring chemiluminescent images.