Order Entry
Spain
ContactUsLinkComponent
Method 551.1 pesticide/herbicide mix (16 components)
Method 551.1 pesticide/herbicide mix (16 components)
Catalog # REST32438
Supplier:  Restek
CAS Number:  
undefined
Method 551.1 pesticide/herbicide mix (16 components)
Catalog # REST32438
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml each in acetone
  • Pk:
    1 mL
  • Description:
    Method 551.1 pesticide/herbicide mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains 16 components, those are alachlor, atrazine, bromacil, cyanazine , endrin, endrin aldehyde, endrin ketone, γ-bhc, heptachlor, heptachlor epoxide, hexachlorobenzene, hexachlorocyclopentadiene, methoxychlor, metolachlor, simazine and trifluralin.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.