Order Entry
Spain
ContactUsLinkComponent
Phenylurea pesticide mixture (8 components)
Phenylurea pesticide mixture (8 components)
Catalog # REST32434
Supplier:  Restek
CAS Number:  
undefined
Phenylurea pesticide mixture (8 components)
Catalog # REST32434
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 μg/ml each in acetonitrile:acetone (90:10)
  • Pk:
    1 mL
  • Description:
    Phenylurea pesticide mixture
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains eight components, those are diflubenzuron, diuron, fluometuron, linuron, propanil, siduron, tebuthiuron and thidiazuron.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.