531.2 carbamate pesticide calibration mixture (11 components)
Catalog # REST32435
Proveedor: Restek
Número CAS:
Especificaciones
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Description:531.2 carbamate pesticide calibration mixture
- Pk Info:1 ml/ampoule
Especificaciones
Acerca de este artículo
This mix contains 11 components.
- Complete set of materials for N-methylcarbamoyloximes and N-methylcarbamates
- New mix satisfies latest update of EPA Method 531.2
- Formulated in acetonitrile for stability and convenience for LC analysis
Components are aldicarb, aldicarb sulphone, aldicarb sulphoxide, carbaryl, carbophuran, 3-hydroxycarbophuran, methiocarb, methomyl, 1-naphthol, oxamyl, propoxur (baygon).
Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.