Order Entry
Spain
ContactUsLinkComponent
Organochlorine pesticide mix AB #3 (20 components)
Organochlorine pesticide mix AB #3 (20 components)
Catalog # REST32415
Proveedor:  Restek
Número CAS:  
undefined
Organochlorine pesticide mix AB #3 (20 components)
Catalog # REST32415
Proveedor:  Restek
Número CAS:  

Especificaciones

  • Concentration:
    2000 µg/ml each in hexane:toluene (1:1)
  • Pk:
    1 mL
  • Description:
    Organochlorine pesticide mix AB #3
  • Pk Info:
    1 ml/ampoule

Especificaciones

Acerca de este artículo

This mix contains 20 components.

Components are aldrin, α-BHC, β-BHC, δ-BHC, γ-BHC, cis-chlordane, trans-chlordane, 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide (isomer B), methoxychlor.

Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.