Organochlorine pesticide mix AB #2 (20 components)
Catalog # REST32292
Supplier: Restek
CAS Number:
Specifications
- Concentration:In hexane:toluene (1:1)
- Pk:1 mL
- Description:Organochlorine pesticide mix AB #2
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains 20 components.
Components are aldrin (8 μg/ml), α-BHC (8 μg/ml), β-BHC (8 μg/ml), δ-BHC (8 μg/ml), γ-BHC (8 μg/ml), cis-chlordane (8 μg/ml), trans-chlordane (8 μg/ml), 4,4'-DDD (16 μg/ml), 4,4'-DDE (16 μg/ml), 4,4'-DDT (16 μg/ml), dieldrin (16 μg/ml), endosulphan I (8 μg/ml), endosulphan II (16 μg/ml), endosulphan sulphate (16 μg/ml), endrin (16 μg/ml), endrin aldehyde (16 μg/ml), endrin ketone (16 μg/ml), heptachlor (8 μg/ml), heptachlor epoxide (isomer B) (8 μg/ml), methoxychlor (8 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.