Order Entry
Spain
ContactUsLinkComponent
8270 calibration mix #4 (22 components)
8270 calibration mix #4 (22 components)
Catalog # REST31621
Supplier:  Restek
CAS Number:  
undefined
8270 calibration mix #4 (22 components)
Catalog # REST31621
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    8270 calibration mix #4
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains: Acetophenone, azobenzene, benzyl alcohol, bis(2-ethylhexyl)phthalate, benzyl butyl phthalate, dibenzofuran, diethylphthalate, dimethylphthalate, di-n-butyl phthalate, di-n-octyl phthalate, 2,4-dinitrotoluene, 2,6-dinitrotoluene, ethyl methanesulfonate, isophorone, isosafrole, methyl methanesulfonate, 1,4-naphthoquinone, nitrobenzene, 4-nitroquinoline-n-oxide, phenacetin, safrole, 1,3,5-trinitrobenzene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.