Order Entry
Spain
ContactUsLinkComponent
524 Internal standard/surrogate mix (3 components)
524 Internal standard/surrogate mix (3 components)
Catalog # REST30201
Supplier:  Restek
CAS Number:  
undefined
524 Internal standard/surrogate mix (3 components)
Catalog # REST30201
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1
  • Description:
    Internal standard/surrogate mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichlorobenzene-d4 (2199-69-1), Fluorobenzene (462-06-6).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.