Order Entry
Spain
ContactUsLinkComponent
 
Calibration check mix, 8270 acid (6 components)
  REST31617
 :  Restek
 :  
undefined
Calibration check mix, 8270 acid (6 components)
  REST31617
 :  Restek
 :  

 

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    Calibration check mix, 8270 acid
  • Pk Info:
    1 ml/ampoule

 

 

Contains: 4-chloro-3-methylphenol, 2,4-dichlorophenol, 2-nitrophenol, pentachlorophenol, phenol, 2,4,6-trichlorophenol.

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.