Order Entry
Spain
ContactUsLinkComponent
L/C VOA Internal standard mix (3 components)
L/C VOA Internal standard mix (3 components)
Catalog # REST30091
Supplier:  Restek
CAS Number:  
undefined
L/C VOA Internal standard mix (3 components)
Catalog # REST30091
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2500 μg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    Internal standard mix, L/C VOA
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: Chlorobenzene-d5 (3114-55-4), 1,4-Dichlorobenzene-d4 (3855-82-1), 1,4-Difluorobenzene (540-36-3).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.