8021 Surrogate mix (2 components)
REST30086
: Restek
:
- Concentration:1500 μg/ml each in P and T methanol
- Pk:1 mL
- Description:Surrogate mix, 8021
- Pk Info:1 ml/ampoule
Mix contains: 2-Bromochlorobenzene (694-80-4), 1,4-Dichlorobutane (110-56-5).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.