Order Entry
Spain
ContactUsLinkComponent
8011 Calibration mix, EDB/DBCP (2 components)
8011 Calibration mix, EDB/DBCP (2 components)
Catalog # REST30062
Supplier:  Restek
CAS Number:  
undefined
8011 Calibration mix, EDB/DBCP (2 components)
Catalog # REST30062
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    Calibration mix - EDB/DBCP
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8), 1,2-Dibromoethane (EDB) (106-93-4).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.