SV calibration mix #6 (18 components)
Catalog # REST31012
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml each in toluene:hexane (1:1)
- Pk:1 mL
- Description:SV calibration mix #6
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
sv calibration mix #6 contains aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc (lindane), 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide (isomer B), methoxychlor.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.