Order Entry
Spain
ContactUsLinkComponent
Residual solvents class 2 mix A (15 components)
Residual solvents class 2 mix A (15 components)
Catalog # REST36271
Supplier:  Restek
CAS Number:  
undefined
Residual solvents class 2 mix A (15 components)
Catalog # REST36271
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In dimethyl sulfoxide
  • Pk:
    1 Ampoul
  • Description:
    Residual solvents class 2 mix A
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains: Acetonitrile, 2,05 mg/ml, chlorobenzene, 1,80 mg/ml, cyclohexane , 19,40 mg/ml, cis-1,2-dichloroethene, 4,70 mg/ml, trans-1,2-dichloroethene, 4,70 mg/ml, 1,4-dioxane, 1,90 mg/ml, ethylbenzene, 1,84 mg/ml, methanol, 15,00 mg/ml, methylcyclohexane, 5,90 mg/ml, methylene chloride (dichloromethane), 3,00 mg/ml, tetrahydrofuran, 3,45 mg/ml, toluene, 4,45 mg/ml, m-xylene, 6,51 mg/ml, o-xylene, 0,98 mg/ml, p-xylene, 1,52 mg/ml.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.