VOA Surrogate spike mix (3 components)
Catalog # REST30004
Supplier: Restek
CAS Number:
Specifications
- Concentration:2500 μg/ml each in P and T methanol
- Pk:1
- Description:VOA surrogate spike mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Mix contains: 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichloroethane-d4 (17060-07-0) and Toluene-d8 (2037-26-5).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.