VOA screening mix #2 (2 components)
Catalog # REST30002
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in P and T methanol
- Pk:1 mL
- Description:VOA screening mix #2
- Pk Info:1 ml/ampoule
Specifications
About this item
Mix contains: (C9) n-Nonane (111-84-2), (C12) n-Dodecane (112-40-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.