Order Entry
Spain
ContactUsLinkComponent
CCME PHC calibration mix (3 components)
CCME PHC calibration mix (3 components)
Catalog # REST31684
Supplier:  Restek
CAS Number:  
undefined
CCME PHC calibration mix (3 components)
Catalog # REST31684
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    5000 µg/ml each in toluene
  • Pk:
    1
  • Description:
    CCME PHC calibration mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This calibration mix consist of three components: (C10) n-Decane, (C16) n-Hexadecane and (C34) n-Tetratriacontane.

  • Meets CCME 2001 petroleum hydrocarbons in soil method—Tier 1
  • Primary reference calibration standards for quantification of four fractions

Shelf life 6 to 36 months.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.