Chlorinated acids by LC, mix B (8 components)
Catalog # REST32430
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in acetonitrile
- Pk:1 mL
- Description:Chlorinated acids by LC, mix B
- Pk Info:1 ml/ampoule
Specifications
About this item
Consists of eight components, those are 2,4-db, 3,5-dichlorobenzoic acid, dinoseb, MCPA, MCPP, 4-nitrophenol, pentachlorophenol and 2,4,5-t.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.