WA VPH marker standard (9 components)
Catalog # REST30450
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in P and T methanol
- Pk:1 mL
- Description:WA VPH marker standard
- Pk Info:1 ml/ampoule
Specifications
About this item
WA VPH marker standard contains (C5) n-pentane, (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, 1-methylnaphthalene, naphthalene, toluene, 1,2,3-trimethylbenzene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.