Florisil cartridge quality control standard mixture, rev. 2 (2 components)
Catalog # REST31677
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in hexane
- Pk:1
- Description:Florisil cartridge quality control standard mixture, rev. 2
- Pk Info:10 ml/ampoule
Specifications
About this item
The mix consists of diesel fuel #2 composite and mineral oil.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.