Order Entry
Spain
ContactUsLinkComponent
Free fatty acids test standard (6 components)
Free fatty acids test standard (6 components)
Catalog # REST35272
Supplier:  Restek
CAS Number:  
undefined
Free fatty acids test standard (6 components)
Catalog # REST35272
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in water
  • Pk:
    1 mL
  • Description:
    Free fatty acids test standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The free fatty acids test standard consists of six components they are (C4:0) butyric acid, acetic acid, isobutyric acid, isovaleric acid, propionic acid and valeric acid.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.