European pharmacopoeia/ICH Class 2 mix C (6 components)
Catalog # REST36231
Supplier: Restek
CAS Number:
Specifications
- Concentration:Prepared in water
- Pk:1
- Description:European pharmacopoeia/ICH Class 2 mixture C
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
The european pharmacopoeia/ICH Class 2 mix C consists of six components, they are 2-ethoxyethanol (160 μg/ml), ethylene glycol (620 μg/ml), formamide (220 μg/ml), 2-methoxyethanol (methyl cellosolve) (50 μg/ml), n-methylpyrrolidone (4,840 μg/m)l and sulpholane (160 μg/m)l.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.