Order Entry
Spain
Orders LinkContactUsLinkComponent
USP <467> calibration mixture #4 (5 components)

USP <467> calibration mixture #4 (5 components)

USP <467> calibration mixture #4, In methanol, 1 ml/ampoule, 1 mL
Catalog # REST36006
Supplier:   Restek
USP <467> calibration mixture #4 (5 components)
Catalog # REST36006
Supplier:  Restek
CAS Number:  
USP <467> calibration mixture #4 (5 components)
USP <467> calibration mixture #4 (5 components)

USP <467> calibration mixture #4, In methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
51,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

The mixture consists of benzene (2 μg/ml), chloroform (60 μg/ml), 1,4-dioxane (380 μg/ml), methylene chloride (dichloromethane) (600 μg/ml) and trichloroethene (80 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.

Specifications

Specifications

  • Concentration:
    In methanol
  • Pk:
    1 mL
  • Description:
    USP <467> calibration mixture #4
  • Pk Info:
    1 ml/ampoule

Specifications