USP calibration mixture #3 (5 components)
Catalog # REST36004
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulphoxide
- Pk:1
- Description:USP <467> calibration mixture #3
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
The mixture consists of benzene (100 μg/ml), chloroform 50 μg/ml, 1,4-dioxane (100 μg/ml), methylene chloride (dichloromethane) (500 μg/ml) and trichloroethene (100 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.