Order Entry
Spain
ContactUsLinkComponent
LC normal phase test mix #1 (4 components)
LC normal phase test mix #1 (4 components)
Catalog # REST35004
Supplier:  Restek
CAS Number:  
undefined
LC normal phase test mix #1 (4 components)
Catalog # REST35004
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In hexane
  • Pk:
    1 mL
  • Description:
    LC normal phase test mixture #1
  • Environmentally Preferable:
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Routine analysis using this mix can assist in determining the need to perform column and/or system maintenance.

The LC normal phase test mix #1 consists of four components they are benzene (1,00 mg/ml), benzaldehyde, (0,04 mg/ml), benzyl alcohol, (3,00 mg/ml) and 4-methoxybenzyl alcohol (2,00 mg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.