Order Entry
Spain
Orders LinkContactUsLinkComponent
4,4'-DDE

4,4'-DDE

4,4'-DDE, 1000 μg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32202
Supplier:   Restek
4,4'-DDE
Catalog # REST32202
Supplier:  Restek
CAS Number:  
4,4'-DDE
4,4'-DDE

4,4'-DDE, 1000 μg/ml in methanol, 1 ml/ampoule, 1

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 μg/ml in methanol
  • Pk:
    1
  • Description:
    4,4'-DDE
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32202
  • Formula:
    C₁₄H₉Cl₅
  • Description:
    4,4'-DDE
  • CAS No.:
    72-55-9

Specifications