4,4'-DDD
4,4'-DDD, 1000 μg/ml in methanol, 1 ml/ampoule, 1
REST32201
: Restek
: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
- Concentration:1000 μg/ml in methanol
- Pk:1
- Description:4,4'-DDD
- Pk Info:1 ml/ampoule
- Cat. No.:REST32201
- Formula:C₁₄H₁₀Cl₄
- Description:4,4'-DDD
- CAS No.:72-54-8



