508.1 calibration mix #2 (11 components)
REST32095
: Restek
:
- Concentration:500 µg/ml each in ethyl acetate
- Pk:1
- Description:508.1 calibration mix #2
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Consists of 11 components, those are chlorobenzilate, cis-chlordane, trans-chlordane, chloroneb, dcpa methyl ester , etridiazole, hexachlorobenzene, cis-permethrin*, trans-permethrin*, propachlor and trifluralin.
11 components, Contains 500 µg/ml total permethrin.
: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
: Exact content of each isomer is listed on certificate of analysis.