Order Entry
Spain
ContactUsLinkComponent
508.1 calibration mix #1 (17 components)
508.1 calibration mix #1 (17 components)
Catalog # REST32094
Supplier:  Restek
CAS Number:  
undefined
508.1 calibration mix #1 (17 components)
Catalog # REST32094
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in ethyl acetate
  • Pk:
    1
  • Description:
    508.1 calibration mix #1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Consists of 17 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc , 4,4'-ddd, 4,4'-dde, 4,4'-ddt, dieldrin, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, heptachlor, heptachlor epoxide and methoxychlor.

17 components

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.