USP Calibration mix #6 (4 components)
Catalog # REST36008
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:USP <467> Calibration mix #6
- Environmentally Preferable:
Specifications
About this item
Mix contains: Chloroform (67-66-3), 60 µg/ml; 1,4-Dioxane (123-91-1), 380 µg/ml; Methylene chloride (dichloromethane) (75-09-2), 600 µg/ml; Trichloroethene (79-01-6), 80 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.