Order Entry
Spain
ContactUsLinkComponent
SV calibration mix #1 (5 components)
SV calibration mix #1 (5 components)
Catalog # REST31007
Supplier:  Restek
CAS Number:  
undefined
SV calibration mix #1 (5 components)
Catalog # REST31007
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    SV calibration mix #1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

SV calibration mix #1 contains benzyl alcohol, 4-chloroaniline, 2-nitroaniline, 3-nitroaniline, 4-nitroaniline.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.