Order Entry
Spain
ContactUsLinkComponent
 
Tetra-n-propyltin internal standard
Catalog # REST31474
Supplier:  Restek
CAS Number:  
Tetra-n-propyltin internal standard
Catalog # REST31474
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methylene chloride
  • Pk:
    1 mL
  • CAS No.:
    2176-98-9
  • Description:
    Tetra-n-propyltin
  • Formula:
    C₁₂H₂₈Sn
  • Description:
    Tetra-n-propyltin
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Complete data pack available for audit compliance.