Butyltin chloride calibration mixture (4 components)
Catalog # REST31472
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Pk:1 mL
- Description:Butyltin chloride calibration mixture
- Pk Info:1 ml/ampoule
Specifications
About this item
This butyltin chloride calibration mixture contain four components: n-Butyltin trichloride, Dibutyltin dichloride, Tetrabutyltin, Tributyltin chloride.
- Complete data pack available for audit compliance