1,3-Dichlorobenzene
REST31443
: Restek
:
- Concentration:1000 µg/m in methanol
- Pk:1 mL
- CAS No.:541-73-1
- Description:1,3-Dichlorobenzene
- Formula:C₆H₄Cl₂
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.