504.1 Calibration mix (3 components)
Catalog # REST30239
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 μg/ml each in P and T methanol
- Pk:1 mL
- Description:Calibration mix, 504,1
- Pk Info:1 ml/ampoule
Specifications
About this item
Mix contains: Chlorobenzene-d5 (3114-55-4); 1,4-Dichlorobenzene-d4 (3855-82-1); Fluorobenzene (462-06-6).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.